ENA394767125 Enamine

4-{cyano[(trimethylsilyl)oxy]methyl}benzonitrile

  • MDL number MFCD16786552
  • Cas Number : 73183-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127788
Availability details for
Quantity
  • Availability
  • Add to Cart