ENA394767143 Enamine

4-oxo-3,4-dihydro-2H-1-benzopyran-6-sulfonyl chloride

  • MDL number MFCD19201154
  • Cas Number : 503470-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127833
Availability details for
Quantity
  • Availability
  • Add to Cart