ENA394767153 Enamine

3-(4-fluorobenzoyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD11607091
  • Cas Number : 304018-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127878
Availability details for
Quantity
  • Availability
  • Add to Cart