ENA394767161 Enamine

7-bromo-1H,2H,3H,4H-pyrido[2,3-b]pyrazin-2-one

  • MDL number MFCD08062978
  • Cas Number : 709652-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127902
Availability details for
Quantity
  • Availability
  • Add to Cart