ENA394767162 Enamine

4-fluoro-1-(2-methoxyethoxy)-2-nitrobenzene

  • MDL number MFCD13689271
  • Cas Number : 1092496-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127903
Availability details for
Quantity
  • Availability
  • Add to Cart