ENA394767164 Enamine

2-acetyl-2,3,4,5-tetrahydro-1H-2-benzazepin-5-one

  • MDL number MFCD24842805
  • Cas Number : 1443981-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127909
Availability details for
Quantity
  • Availability
  • Add to Cart