ENA394767171 Enamine

8-fluoro-2-hydroxy-6-methylquinoline-4-carboxylic acid

  • MDL number MFCD16167598
  • Cas Number : 1248399-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 337...339
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127954
Availability details for
Quantity
  • Availability
  • Add to Cart