ENA394767174 Enamine

2-chloro-6-(methylsulfanyl)benzonitrile

  • MDL number MFCD16780144
  • Cas Number : 51271-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127995

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127995
Availability details for
Quantity
  • Availability
  • Add to Cart