ENA394767188 Enamine

tert-butyl 4-phenyl-1,2,3,6-tetrahydropyridine-1-carboxylate

  • MDL number MFCD11977149
  • Cas Number : 186347-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-128015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128015
Availability details for
Quantity
  • Availability
  • Add to Cart