ENA394767197 Enamine

3-methyl-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD03447190
  • Cas Number : 7114-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128049
Availability details for
Quantity
  • Availability
  • Add to Cart