ENA394767201 Enamine

2,5-dimethyl-4-nitrobenzonitrile

  • MDL number MFCD00024285
  • Cas Number : 73713-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128064

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-128064
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart