ENA394767206 Enamine

4-hydroxy-N-phenylbenzamide

  • MDL number MFCD01017298
  • Cas Number : 14121-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128091
Availability details for
Quantity
  • Availability
  • Add to Cart