ENA394767207 Enamine

1-(2,4-difluorophenyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD11543140
  • Cas Number : 1038333-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128092
Availability details for
Quantity
  • Availability
  • Add to Cart