ENA394767214 Enamine

2-(3-aminophenyl)ethyl acetate

  • MDL number MFCD20697011
  • Cas Number : 690995-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128100
Availability details for
Quantity
  • Availability
  • Add to Cart