ENA394767219 Enamine

N-[2-(trifluoromethyl)phenyl]hydrazinecarbothioamide

  • MDL number MFCD00060569
  • Cas Number : 38901-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03799
Availability details for
Quantity
  • Availability
  • Add to Cart