ENA394767222 Enamine

2,2-dimethylpent-4-yn-1-ol

  • MDL number MFCD19441316
  • Cas Number : 107540-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128110
Availability details for
Quantity
  • Availability
  • Add to Cart