ENA394767227 Enamine

(2,5-dichlorophenyl)(oxolan-3-yl)methanamine

  • MDL number MFCD14610397
  • Cas Number : 1248787-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128115
Availability details for
Quantity
  • Availability
  • Add to Cart