ENA394767228 Enamine

N-[2-(furan-2-yl)ethyl]acetamide

  • MDL number MFCD11727033
  • Cas Number : 861058-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128116

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128116
Availability details for
Quantity
  • Availability
  • Add to Cart