ENA394767232 Enamine

2-(3-hydroxyphenyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12107712
  • Cas Number : 1155592-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128121
Availability details for
Quantity
  • Availability
  • Add to Cart