ENA394767236 Enamine

(1R)-1-(3-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD11101211
  • Cas Number : 1167414-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128127
Availability details for
Quantity
  • Availability
  • Add to Cart