ENA394767246 Enamine

ethyl 7-nitro-1,3-dioxaindane-5-carboxylate

  • MDL number MFCD12412148
  • Cas Number : 1156940-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128154
Availability details for
Quantity
  • Availability
  • Add to Cart