ENA394767248 Enamine

6-bromo-2-oxo-2,3-dihydro-1H-indole-3-carbaldehyde

  • MDL number MFCD20528307
  • Cas Number : 99365-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128156

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128156
Availability details for
Quantity
  • Availability
  • Add to Cart