ENA394767266 Enamine

ethyl 2-methyl-3-(4-methylphenyl)prop-2-enoate

  • MDL number MFCD24842830
  • Cas Number : 274907-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128187
Availability details for
Quantity
  • Availability
  • Add to Cart