ENA394767267 Enamine

2-methyl-3-(4-methylphenyl)propanoic acid

  • MDL number MFCD00796497
  • Cas Number : 1012-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128188
Availability details for
Quantity
  • Availability
  • Add to Cart