ENA394767271 Enamine

methyl 5-(chlorosulfonyl)-2-iodobenzoate

  • MDL number MFCD12169464
  • Cas Number : 1065102-81-9
  • Molecular Weight : 360.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-(chlorosulfonyl)-2-iodobenzoate
Inchi Code 1S/C8H6ClIO4S/c1-14-8(11)6-4-5(15(9,12)13)2-3-7(6)10/h2-4H,1H3
Inchi Key YZSOCOKTNYPOFU-UHFFFAOYSA-N

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128192

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart