ENA394767277 Enamine

2-hydroxy-1-(4-methoxyphenyl)ethan-1-one

  • MDL number MFCD00004668
  • Cas Number : 4136-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128204
Availability details for
Quantity
  • Availability
  • Add to Cart