ENA394767282 Enamine

6-acetyl-1H,2H,3H-thieno[2,3-b][1,4]thiazin-2-one

  • MDL number MFCD00219772
  • Cas Number : 151095-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128211

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128211
Availability details for
Quantity
  • Availability
  • Add to Cart