ENA394767285 Enamine

4-chloro-2-ethoxybenzoic acid

  • MDL number MFCD14646682
  • Cas Number : 1195264-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128214

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128214
Availability details for
Quantity
  • Availability
  • Add to Cart