ENA394767286 Enamine

methyl 3-[2,2,2-trifluoro-N-(propan-2-yl)acetamido]propanoate

  • MDL number MFCD24842837
  • Cas Number : 1443981-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128215
Availability details for
Quantity
  • Availability
  • Add to Cart