ENA394767289 Enamine

ethyl 2-methyl-3-(4-methylphenyl)propanoate

  • MDL number MFCD22987365
  • Cas Number : 859191-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128218
Availability details for
Quantity
  • Availability
  • Add to Cart