ENA394767293 Enamine

3-{[(pyrazin-2-yl)amino]methyl}benzonitrile

  • MDL number MFCD12762481
  • Cas Number : 1179134-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128222
Availability details for
Quantity
  • Availability
  • Add to Cart