ENA394767296 Enamine

2-(pyridin-2-yl)-1,3-thiazol-4-amine

  • MDL number MFCD12033358
  • Cas Number : 93209-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128225
Availability details for
Quantity
  • Availability
  • Add to Cart