ENA394767298 Enamine

3-methoxy-2-methylspiro[3.3]heptan-1-amine hydrochloride

  • MDL number MFCD24842838
  • Cas Number : 1443979-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128227
Availability details for
Quantity
  • Availability
  • Add to Cart