ENA394767301 Enamine

4-(benzylcarbamoyl)butanoic acid

  • MDL number MFCD00029858
  • Cas Number : 42856-45-1
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128230
Availability details for
Quantity
  • Availability
  • Add to Cart