ENA394767323 Enamine

4-bromo-2,5-dimethylphenol

  • MDL number MFCD00029746
  • Cas Number : 85223-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128262
Availability details for
Quantity
  • Availability
  • Add to Cart