ENA394767329 Enamine

[(4-methoxy-1H-indol-3-yl)methyl]dimethylamine

  • MDL number MFCD08234941
  • Cas Number : 52335-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128270
Availability details for
Quantity
  • Availability
  • Add to Cart