ENA394767334 Enamine

1-(bromomethyl)-1-(prop-2-yn-1-yl)cyclobutane

  • MDL number MFCD20636507
  • Cas Number : 950577-94-3
  • Molecular Weight : 187.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(bromomethyl)-1-(2-propynyl)cyclobutane
Inchi Code 1S/C8H11Br/c1-2-4-8(7-9)5-3-6-8/h1H,3-7H2
Inchi Key UMJDSEZANNCYRF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128275

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-128275
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart