ENA394767335 Enamine

4-chloro-1,3-thiazole-5-carbonitrile

  • MDL number MFCD18822128
  • Cas Number : 145383-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128276
Availability details for
Quantity
  • Availability
  • Add to Cart