ENA394767336 Enamine

ethyl [(2,2-difluoroethyl)carbamoyl]formate

  • MDL number MFCD24202781
  • Cas Number : 1380394-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128280
Availability details for
Quantity
  • Availability
  • Add to Cart