ENA394767340 Enamine

5-(bromomethyl)-2,3-dihydro-1H-indene

  • MDL number MFCD09261154
  • Cas Number : 501649-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128298
Availability details for
Quantity
  • Availability
  • Add to Cart