ENA394767343 Enamine

4-methyl-2-oxo-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD24842849
  • Cas Number : 1443980-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128302
Availability details for
Quantity
  • Availability
  • Add to Cart