ENA394767352 Enamine

3-cyclopropylquinoline

  • MDL number MFCD18086939
  • Cas Number : 485402-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128313
Availability details for
Quantity
  • Availability
  • Add to Cart