ENA394767353 Enamine

4-ethyloxane-4-carboxylic acid

  • MDL number MFCD12406612
  • Cas Number : 1158759-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128314
Availability details for
Quantity
  • Availability
  • Add to Cart