ENA394767355 Enamine

5,6,7,8-tetrahydropteridine

  • MDL number MFCD01741597
  • Cas Number : 10593-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-147
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128317
Availability details for
Quantity
  • Availability
  • Add to Cart