ENA394767358 Enamine

(2-hydrazinylidene-1,2-diphenylethylidene)hydrazine

  • Cas Number : 4702-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06416
Availability details for
Quantity
  • Availability
  • Add to Cart