ENA394767360 Enamine

[2-(4-chlorophenyl)ethyl](methyl)amine hydrochloride

  • MDL number MFCD10565660
  • Cas Number : 102236-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128339
Availability details for
Quantity
  • Availability
  • Add to Cart