ENA394767364 Enamine

methyl 1-bromocyclopropane-1-carboxylate

  • MDL number MFCD12922645
  • Cas Number : 96999-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 45
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128369

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-128369
pictograms GHS02,GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart