ENA394767365 Enamine

1-cyclohexylidenepropan-2-one

  • MDL number MFCD00462181
  • Cas Number : 874-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128371
Availability details for
Quantity
  • Availability
  • Add to Cart