ENA394767371 Enamine

1-(2,6-difluorophenyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD11202305
  • Cas Number : 1038303-08-0
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128378
Availability details for
Quantity
  • Availability
  • Add to Cart