ENA394767374 Enamine

1-(2-nitroprop-1-en-1-yl)-2-(trifluoromethyl)benzene

  • MDL number MFCD01317566
  • Cas Number : 781-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128382
Availability details for
Quantity
  • Availability
  • Add to Cart